HyFlex® 11-510 Cut Protection Gloves with Plaiting on Palm and Back, Ansell
Supplier: Ansell Healthcare
205743 205744 205745 205746 205747 205748
89171-376CS 3380.63 CAD
89171-376 89171-378 89171-380 89171-382 89171-384 89171-386
HyFlex® 11-510 Cut Protection Gloves with Plaiting on Palm and Back, Ansell
Gloves
Designed to provide cut protection in applications where abrasion resistance and dexterity are a 'must have', these gloves protect hands from abrasion in the palm area as well as the back of the hand, providing more confidence when working in restrictive spaces.
  • Excellent resistance to cuts and abrasions
  • Provide confidence in restrictive work spaces
  • Silicone-free
  • Latex-free
  • Minimal shrinking when laundered
Knitted with DuPont™ Kevlar® yarn, gloves offer flexibility and comfort along with ANSI Level 2 cut protection.
Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
Use in the aerospace, automotive and transportation, construction, metal fabrication, plastic extrusion, public utilities, and white goods industries. Uses include carpentry; pipe fitting; final assembly; glass installation; maintenance operations; sheet metals and metalwork; small parts stamping; small scale metal stamping and assembly; and wiring.
The ComfortFlex cuff provides additional protection as well as a soft feel around the sensitive wrist area. Features DuPont™ Kevlar® stretch liner with spandex and patented Nylon and Kevlar® zonal plaiting on palm and back. Black foam nitrile ensures a sure grip in both light oil and dry applications. Gloves are silicone- and latex-free. Machine launderable with minimal shrinking and good recovery for use again and again.
Certifications: ANSI Level 2 cut protection; ANSI Abrasion Level 4.
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags per case.