BP5MS GC Capillary Column, SGE
Supplier: Trajan Scientific and Medical
054301 054310 054308
89232-102EA 1335.05 CAD
89232-102 89232-104 10799-664
BP5MS GC Capillary Column, SGE
Chromatography Columns
Innovative phase technologies and fused silica production capabilities provide end-to-end separation solutions for applications such as ultra trace analyses, pesticides and herbicides, hydrocarbons, solvents, phenols, amines, GC/MS and other specific detector applications.
  • For your 5% MS analysis
  • Optimized silphenylene for general purpose MS analyses
  • Low bleed
  • Great inertness
  • High temperature
  • Non-polar
Operating temperature from –40 to 330/350 °C.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These 5% Phenyl Polysilphenylene-siloxane columns are suitable replacements for DB-5ms, ZB-5ms, Rxi-5Sil, MS, VF-5ms, CP-Sil 8 CB.