BP10 GC Capillary Column, SGE
Supplier: Trajan Scientific and Medical
054256 054259 054270 054283 054280 054268 054253
14225-926EA 1145.55 CAD
14225-926 14225-932 14225-942 14225-946 60361-886 60361-892 60361-900
BP10 GC Capillary Column, SGE
Chromatography Columns
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
  • Used for organochlorine pesticides analysis
  • Bonded and cross-linked
  • Highly inert
  • Low bleed
  • Able to be solvent rinsed
These columns can be used with environmental analysis (EPA methods 608, 8081), pesticides/herbicides., drugs of abuse and pharmaceuticals.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These 14% cyanopropylphenyl polysiloxaneMore Product Information