Refrigerated Incubators, 7001 Series, Caron Products

Supplier: Caron Products
7001-10-1 7001-10-2 7001-10-3 7001-25-1 7001-25-2 7001-25-3 7001-33-1 7001-33-2 7001-33-3
10216-746EA 39371.97 CAD
10216-746 10216-748 10216-750 10216-752 10216-754 10216-756 10216-758 10216-760 10216-762
Refrigerated Incubators, 7001 Series, Caron Products
Incubators Cooled Incubators
Caron’s Refrigerated Incubators precisely and tightly control temperature conditions from 5 °C to 70 °C with optional operation down to 0 °C, making them the ultimate choice for refrigerated storage.

Our earth-friendly technologies contribute to cost savings without compromising performance and 115V units will plug into a standard laboratory electrical outlet. The precision controlled temperature is excellent for reliable, repeatable results. Key standard features include: touchscreen user interface; built-in preventative maintenance; gROD Refrigeration on Demand control system; tool-lessMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable Energy Efficient Energy Efficient
Temperature uniformity ±0.3°C ±0.3°C
Temperature sensor 3-wire RTD 3-wire RTD
Temperature range 5…70 °C 5…70 °C
Temperature control ±0.1°C ±0.1°C
Shelf dimensions 57.2W x 64.5D cm (22.5 x 25.4") 74.3W x 62.1D cm (29.25 x 24.45")

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR