BP20 GC Capillary Column, SGE
Supplier: Trajan Scientific and Medical
054405 054415 054438 054439 054440 054445 0544515 054456 054457 054458 054444 054451 054450 054448 054443 054442 054437 054436 054434 054433 054431 054428 054427 054425 054424 054422 054421
14225-960EA 806.94 CAD
14225-960 14225-962 14225-964 14225-966 14225-968 14225-970 14225-972 14225-976 14225-978 14225-980 14233-458 60361-830 60361-832 60361-836 60361-840 60361-842 60361-844 60361-846 60361-850 60361-852 60361-856 60361-862 60361-864 60361-868 60361-870 60361-874 60361-876
BP20 GC Capillary Column, SGE
Chromatography Columns
More than five decades of innovative phase technologies and unique fused silica production capabilities together provide end-to-end separation solutions for all applications.
  • Industry standard wax column
  • Bonded and cross-linked
  • Able to be solvent rinsed
These columns can be used with alcohol, free acids, FAME's, aromatics, solvents and essential oils.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: Polyethylene glycol phase columns are suitable replacements for DB-Wax, HP-20M, Supelcowax 10, CB-Wax, Stabilwax, Carbowax, HP-Innowax, Rtx-WAX, PE-WAX, RH-WAX, ZB-WAX and TRWAX.