BP21 GC Capillary Column, SGE
Supplier: Trajan Scientific and Medical
054465 054466 054471 054472 054477 054478 054474 054469 054468 054462 054480
14225-984CS 729 CAD
14225-984 14225-986 14225-990 14225-992 14225-996 14225-998 60361-814 60361-818 60361-820 60361-826 76409-330
BP21 GC Capillary Column, SGE
Chromatography Columns
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
  • Polar phase
  • Ideal for low molecular weight acids
  • Able to be solvent rinsed
  • Bonded and cross-linked
These columns can be used with volatile free acids, FAMEs, alcohols, aldehydes, acrylates and ketones.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These nitroterephthalic acid modified PEG columns are a suitable replacement for DB-FFAP, HP-FFAP, Stabilwax-DA and CPWax-58CB.