VWR® Traceable® Dual Refrigerator/Freezer Thermometers
Supplier: VWR International
10208-394 10208-396
10208-394EA 392.61 CAD
10208-394 10208-396
VWR® Traceable® Dual Refrigerator/Freezer Thermometers
Thermometers Digital Thermometers
Traceable® Dual thermometer displays the temperatures of two separate probes in °C and °F.
  • Easy to operate
  • Resilient against chemicals and impacts
  • Audible and visual alarms
Allows putting one probe in the freezer and another in the refrigerator (or measure two areas of a refrigerator)
High-impact, chemical-resistant ABS plastic case is 3-1/2 x 1-3/4 x 1 inches. Weight is 3-2/3 ounces.
Range is -58 to 158°F and -50 to 70°C with a resolution of 0.1° and an accuracy of ±1°C. Three and a half-foot micro-cable permits refrigerator doors to close on it. Probe is sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about incidental contact with food or drinking water. A unit with Bullet™ probes (without bottles) is also available.
Certifications: Traceable to NIST for Accuracy–An individually-numbered Traceable® Certificate is provided which assures accuracy from VWR® ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Ordering information: Supplied with flip open stand. Velcro® and magnetic strips are provided for mounting of both thermometer and bottle sensor. Supplied with a Traceable® Certificate and AAA battery for continuous, 1-year always-on monitoring.