HT5 GC Capillary Columns, SGE
Supplier: Trajan Scientific and Medical
054633 054634 054667 054672 054670 054668 054658 054657 054655 054642 054641 054632 054631
14226-002EA 736.36 CAD
14226-002 14226-004 14226-006 60361-736 60361-740 60361-742 60361-754 60361-756 60361-758 60361-766 60361-768 60361-774 60361-776
HT5 GC Capillary Columns, SGE
Chromatography Columns
More than five decades of innovative phase technologies and unique fused silica production capabilities, together provide end-to-end separation solutions for all applications.
  • Ultra high temperature columns
  • Unique phases
  • Ideal for simulated distillation applications (petroleum industry)
  • Bonded and cross-linked
  • Able to be solvent rinsed
These columns can be used in simulated distillation, general hydrocarbon profiles, pesticides/herbicides and GC/MS applications.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: The 5% phenyl polycarborane-siloxane columns are a suitable replacement for MXT-1 Sim Dist, HT-Sim, DistCB and MXT-500.