Digital 'Pocket' Refractometer, Model PAL-RI, ATAGO®
Supplier: Atago
3850 3852
97015-894EA 1358.52 CAD
97015-894 97015-896
Digital 'Pocket' Refractometer, Model PAL-RI, ATAGO®
Refractometers Handheld Refractometers
Model PAL-RI is designed for measuring Refractive Index, the measurement will be displayed continuously like an electric newsboard once the sample has been placed
The device features IP65-rated dustproof and water-resistant protection. A sample volume of 0.3mL is required for measurement results.
Ordering information: One-year manufacturer’s warranty. Supplied with a storage case and two AAA batteries. A calibration certificate is available. Contact your Avantor representative for information.
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambient temperature 10...40 °C (50...104 °F) 10...40 °C (50...104 °F)
International protection class IP65 IP65
Measurement temperature range 5 to 45°C (41 to 113°F) 10 to 75°C (50 to 167°F)
Measurement time 3 seconds 3 seconds
Resolution 0.0001 nD 0.0001 nD