Combined Ion Selective Electrodes, 800 Series, WTW
Supplier: WTW
106666 106661 106663 106665 106667
80085-282EA 3349.76 CAD
80085-282 80085-284 80085-286 80085-290 80085-292 80085-294 80085-296 80085-298 80085-300 80085-304 97042-882 97042-884 97042-886 97042-888
Combined Ion Selective Electrodes, 800 Series, WTW
Electrodes ISE Electrodes
These easy-to-use, robust, high quality ion-selective electrodes are ideal for field applications.
  • Combined with built-in reference electrodes which make them particularly suitable for measurement of small volume samples
Combined with built-in reference electrodes which make them particularly suitable for measurement of small volume samples.
Ordering information: These ion selective electrodes will require additional electrolyte and ion strength adjustment solutions (ISA) as well as freshly prepared standards (see ISA solutions).