Prinzing® Wall Switch Lockout, Brady Worldwide®
Supplier: Brady
WSLO
CA56606-532EA 11.99 CAD
CA56606-532
Prinzing® Wall Switch Lockout, Brady Worldwide®
Security Locks
Wall Switch Lockouts may be used with most wall-mounted switches and are simple to install using existing plate screws
Made of clear polycarbonate with a patented non-hinge design. Padlock sold separately.