Flocked Swab for Rapid Testing, Starplex Scientific
Supplier: Starplex Scientific
SP132-FL SP132-FL
CA89130-578CS 2300.2 CAD
CA89130-578 89130-578
Flocked Swab for Rapid Testing, Starplex Scientific
Swabs Swabs for Sampling
Liquid Stuart's media in the durable 10 mL vial and swab retaining cap packaged with a standard flocked swab is ideal for various rapid testing methodology and with liquid sampling instrumentation.