Breaker Lockout Pouch, Brady Worldwide®
Supplier: Brady
65405 65405
CA12222-704EA 239.55 CAD
CA12222-704 12222-704
Breaker Lockout Pouch, Brady Worldwide®
Bags Pouches and Belt Cases
Lightweight, portable pouch contains lockout devices to secure electrical equipment.
  • Pouch is equipped with metal loops for hanging on a belt
Each pouch contains one Small Brady Plug Lockout, three Wall Switch Lockouts, three Universal Multi-Pole Lockouts, three 277 Volt 'No Hole' Lockouts, two 480/600 Volt 'No Hole' Lockouts, one general-use Cleat 277, and one general-use Cleat 480/600.