Trajan, Capillary Electrophoresis Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
062801 062803 062813 062823
14226-226EA 84.53 CAD
14226-226 14226-228 14226-230 14226-232
Trajan, Capillary Electrophoresis Tubing, Trajan Scientific and Medical
Tubing Transfer Tubing
This specialty tubing has been developed through a complete understanding of the chemistry and properties of different types of glass and polymers.
  • Manufactured from precision bore fused silica tubing
  • Hydrophobic and hydrophilic surface treatments
  • Wide pH range
  • Column-to-column reproducibility
It can be used uncoated, or coated in the laboratory.
Certifications: RoHS compliant per Directive 2011/65/EU.