Steel Padlock Labels, Brady
Supplier: Brady Worldwide
50280 50281 50283
CA89007-202PK 13.54 CAD
CA89007-202 CA89007-204 CA89007-206
Steel Padlock Labels, Brady
Labels
Steel padlock labels send a clear message to workers when a machine is down for service or maintenance.
  • Ideal for use in cold storage warehouses
  • Labels accept pencil, pen or marker
  • Labels are available in English, Spanish or French
  • Service temperature range: −50 to 200 °F
Labels are made of durable vinyl and come with overlaminate strips for enhanced durability. This flexible vinyl film is computer printed and has a cold temperature applicable pressure sensitive adhesive.