Combination Lockout Duffel, Brady Worldwide®
Supplier: Brady
99689 99691 99689 99691 99690
CA89008-140KT 669.3 CAD
CA89008-140 CA89008-142 89008-143 89008-139 CA89008-138
Combination Lockout Duffel, Brady Worldwide®
Security Locks
The combination lockout duffel provides everything needed to lockout the most common mechanical and electrical control points
Extra electrical lockout devices are included. Kit components include; one Mini Cable Lockout, one Large Universal Valve Lockout with Blocking Arm, one 120 V Snap-On Breaker Lockout, one 120/277 V Clamp-On Breaker Lockout, one 480/600 V Clamp-On Breaker Lockout, one Universal Multi-Pole Breaker Lockout, one Oversized Breaker Lockout, one Cleat for 120/277 V Clamp-On Lockout, one Cleat for 480/600 V Clamp-On Lockout, one 3-In-1 Plug Lockout, one Wall Switch Lockout, one 1–6.5" Adjustable Gate Valve Lockout, one 1 to 2.5" Gate Valve Lockout, one 0.25 to 1" Ball Valve Lockout, one Lockout Belt Pouch, one Plug Valve Lockout for stem diameters up to 0.875", one Plug Valve Lockout for stem diameters 0.281 to 1.375", and one Lockout Duffel Bag.