Prinzing® Economy Lockout Kit, Brady Worldwide®
Supplier: Avantor
CA89008-186EA 193.45 CAD
CA89008-186 CA89008-188
Prinzing® Economy Lockout Kit, Brady Worldwide®
Security Locks
The economy lockout kit provides nine components to ensure equipment is either de-energized or locked out in accordance with OSHA regulation 1910.47.
Delivery information: Kit components include; three 1.5" Steel Lockout Hasps, three Blue Prinzing® Padlocks, fourteen 2 Padlock Identifier Stickers, one 10 Pack Lockout Tags With Ties, three Single Pole Breaker Lockouts, two Multi-Pole Breaker Lockouts, one Large Plug Lockout, one Lockout Training Booklet, and one Rugged Polyethylene Carrying Case (7H×12W×5"D).