Valve Lockout Pouch Kit, Brady Worldwide®
Supplier: Avantor
CA89008-120KT 204.57 CAD
CA89008-120 CA89008-122 CA89008-124
Valve Lockout Pouch Kit, Brady Worldwide®
Security Locks
The economical valve lockout pouch kit is perfect for locking out small gate and ball valves.
Delivery information: Kit components includes 1 to 6.5" Adjustable Gate Valve Lockout, 0.5 to 1.25" Small Ball Valve Lockouts (2), 1 Plug Valve Lockout for stem diameters up to 0.875", 1 Plug Valve Lockout for stem diameters 0.281 to 1.375" and 1 Large Lockout Pouch.