Electrical Plug Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
65675 65674
CA12222-764EA 29.26 CAD
CA12222-764 CA12222-766
Electrical Plug Lockouts, Brady Worldwide®
Security Locks
Electrical plug lockouts are a simple solution for electric equipment lockout procedures. They prevent electrical plugs from being inserted into a wall outlet.
  • Accommodate a large variety of plug shapes and sizes
  • Constructed of rugged polypropylene
  • Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs