Wall Switch Lockouts, Brady Worldwide®
Supplier: Brady Worldwide
65696 65392
CA12222-752PK 74.5 CAD
CA12222-752 CA12222-760
Wall Switch Lockouts, Brady Worldwide®
Security Locks
Wall switch lockouts are designed for use with most wall-mounted switches and are simple to install using existing screws
Designed with a high visibility red finish, wall switch lockouts can be used to lock switches "on" or "off". Padlock sold separately.