10-Lock Board, Brady Worldwide®
Supplier: Brady Worldwide
50990
CA89007-230EA 186.07 CAD
CA89007-230 CA89007-254 CA89007-256
10-Lock Board, Brady Worldwide®
Security Locks
A successful lockout program begins with the right devices stored in the right places
Brady® lockout station boards are the perfect solution to get started. Boards are made of 3/16" thick, bright yellow acrylic with acrylic lock rails. Tag pockets are made of clear polycarbonate and will hold tags up to 3" wide. Tags, padlocks, and hasps not included.