Snap Cap™ Gas Cylinder Lockout, Brady Worldwide®
Supplier: Brady Worldwide
95137 95138 95139
CA89007-920EA 213.57 CAD
CA89007-920 CA89007-922 CA89007-924
Snap Cap™ Gas Cylinder Lockout, Brady Worldwide®
Security Locks
Designed to fit most North American high and low pressure gas cylinders, the Snap Cap™ device denies access to the main cylinder control and regulator attachment
Constructed of welded steel and powder-coated for many years of use.