Wrist Action Variable Speed Shaker, Burrell Scientific
Supplier: Burrell Scientific
075-795-04-19 075-795-24-19
10161-166EA 14343.54 CAD
10161-166 10799-402
Wrist Action Variable Speed Shaker, Burrell Scientific
Shakers Rotating/Rocking Shakers
Wrist Action® Variable Speed Shaker has a variable speed motor which will oscillate from 0 450 Osc/Min.This Shaker has a built in timer to operate from 1 to 60 minutes or can be set to continuous operation.
  • With the appropriate clamps and supports it can hold a variety of flask, funnels, and specimen jar
  • Hold up to ten 125 mL, eight 250/300 mL, or six 500 mL flask
The Wrist Action® Shaker is designed to maintain a constant swirling action as long as required for almost any laboratory application. The adjustment arm controls the amplitude and agitationMore Product Information