Vacuum Pump System with Wireless Remote, Model SC920, KNF Neuberger
Supplier: KNF
USC920
97014-790EA 21652.23 CAD
97014-790
Vacuum Pump System with Wireless Remote, Model SC920, KNF Neuberger
Pumps Vacuum Pumps
Delivers precise vacuum with easy, safe control and monitoring from anywhere in the lab.
  • Intuitive interface with touch controls and rotation knob
  • Menus include unit of measurement, language, mode, parameters
  • Graphic display shows process variables
  • Evacuation (regulated pumping)
  • Controlled emptying of a chamber
  • Pressure Control; system controls pressure to set value (constant pressure)
  • Automatic; system automatically finds vapor pressure of sample and adjusts process pressure accordingly
  • Individual Pressure Function; system controls process according to user-defined pressure curve
  • REPEAT function allows successive repetitions of programmed curve
  • Manual process control can be selected at any time during an active process
  • PC control Windows-based software (included) enables control via external PC
  • Software supports display of pressure curves as a chart
  • Setpoints can be saved
  • Data can be exported to spreadsheets or text files
  • USB interface
Oil-free vacuum system supplies clean, reliable vacuum source without hassles of handling pump oils or tainted aspiration water. Four operating modes make this a fast, versatile device for a wide range of pumping applications. Intelligent control system monitors actual pressure; system compensates for fluctuation. Low-speed start. Patented diaphragm stabilization system ensures high speeds at low pressures for faster processing times. System stops when setpoint pressure is achieved; cycles ON/OFF intermittently to stabilize pressure. Chemical-resistant gas carrying parts. Quiet operation.