Angled Two Neck Round Bottom Flask, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
294990-0010 294990-0025 294990-0050 605020-0210 605020-1524 605020-2924 295000-0025 295010-0050 295010-0100 295010-0250 605020-0624 605020-1024 605020-1824
46200-650EA 481.47 CAD
46200-650 KT294990-0010 KT294990-0025 KT294990-0050 KT294990-0100 KT294990-0250 KT294990-0500 KT605020-0210 KT605020-0610 KT605020-1424 KT605020-1524 KT605020-2624 KT605020-2924 KT294990-0015 KT295000-0025 KT295010-0025 KT295010-0050 KT295010-0100 KT295010-0250 KT605020-0224 KT605020-0624 KT605020-1024 KT605020-1824
Angled Two Neck Round Bottom Flask, Kimble Chase
Flasks Round Bottom Flasks
Distilling round bottom flask with angled side neck
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-neck flask with Standard Taper outer joints. Side neck is angled.