Lab-Sorb™ for Liquid Disposal, Diversified Biotech
Supplier: Diversified Biotech
SORB-1000 SORB-2000 SORB-2100 SORB-2200 SORB-2300 SORB-3000 SORB-3100 SORB-3200
10141-806EA 103.25 CAD
10141-806 10141-808 10141-810 10141-812 10141-814 10141-816 10141-818 10141-820
Lab-Sorb™ for Liquid Disposal, Diversified Biotech
Spill Control Materials
  • Converts liquid waste to solid waste in seconds
  • Lab-Sorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
  • One 16 oz. jar can absorb up to 50 liters of liquid