VWR® E Balances
Supplier: VWR International
10204-990 10204-992 10204-994 10204-996 10204-998 10205-000
10204-990EA 1896.98 CAD
10204-990 10204-992 10204-994 10204-996 10204-998 10205-000
VWR® E Balances
Balances and Scales
Entry-level balances designed for basic weighing needs.
  • Economical and versatile
  • Large, backlit display
  • Simple, five-button operation
  • 5-year warranty
Sturdy construction and ease-of-use make the VWR® E-Series balance the ideal choice for the laboratory.
VWR-10001E and VWR-20001E feature an internal rechargeable battery, all other models can be powered by 6xAA batteries. RS232 interface allows connection to VWR® Dot Matrix Printer (cat. no. 10799-132).