VWR® A Balances
Supplier: VWR International
10204-966 10204-968 10204-970 76300-142 76300-144 76300-146 76300-152 76446-810 76446-812 76446-814 76446-816 76446-818 76446-820 76446-822
10204-966EA 3404.71 CAD
10204-966 10204-968 10204-970 76300-142 76300-144 76300-146 76300-152 76446-810 76446-812 76446-814 76446-816 76446-818 76446-820 76446-822
VWR® A Balances
Balances and Scales
VWR® A-Series balances incorporate state-of-the-art features for advanced weighing applications and documentation.
  • State-of-the-art features for advanced weighing and documentation
  • Internal calibration
  • Multiple weigh modes
  • 2×RS-232, USB (A and B), and Wi-Fi communication interfaces
  • Easy-to-read, adjustable, backlit display
  • Tool-free assembly and diassembly of the 3-door draft shield on analytical models
  • 5 year warranty
The VWR® A-Series balances are the answer for organizations demanding accuracy and performance at an economical price. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-Series provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its unsurpassed 5-year warranty, the VWR® A-Series balance will provide reliable results for years to come.