Human CD14+ Monocytes (hMoCD14+-PB), PromoCell
Supplier: Promocell
C-12909 C-14110
10175-182EA 2087.72 CAD
10175-182 10175-222
Human CD14+ Monocytes (hMoCD14+-PB), PromoCell
Cells
PromoCell Human CD14+ Monocytes are isolated from fresh peripheral blood. After isolation of the ultra-pure mononuclear cell fraction by proprietary methods, the CD14+ Monocytes are isolated by positive selection with > 95% purity.
In addition, hMoCD14+-PB are available which are pre-screened for their differentiation capacity into dendritic cells.
This product can be used in the following applications. Dendritic Cell Research, (Auto-)Immunity, Infection biology
Ordering information: Human CD14+ cells can be orderd as cryopreserved cells. Additional cell pellets stored in RNAlater® are available.