Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM), PromoCell
Supplier: Promocell
C-12974 C-14090
10172-152EA 1925.45 CAD
10172-152 10172-154 10172-268
Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM), PromoCell
Cells
PromoCell hMSC-BM are harvested from normal human bone marrow. The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, osteoblasts, and neuronal cells.
Mesenchymal Stem Cells (MSC), also termed Mesenchymal Stromal Cells, are multipotent cells that can differentiate into a variety of cell types and have the capacity for self renewal
This product can be used in the following applications. Stem cell research, Regenerative medicine research, Tissue engineering studies, Developmental experiments.
MSC have been shown to differentiate in vitro or in vivo into adipocytes, chondrocytes, osteoblasts, myocytes, neurons, hepatocytes, and pancreatic islet cells. Optimized PromoCell media are available to support both the growth of MSC and their differentiation into several different lineages. Recent experiments suggest that differentiation capabilities into diverse cell types vary between MSC of different origin
Ordering information: Human Mesenchymal Stem Cells from Bone Marrow can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.