Human Uterine Smooth Muscle Cells (HUtSMC), PromoCell
Supplier: Promocell
C-12575 C-14059
10172-116EA 1927.24 CAD
10172-116 10172-236 10175-144
Human Uterine Smooth Muscle Cells (HUtSMC), PromoCell
Cells
Primary Human Uterine Smooth Muscle Cells (HUtSMC) are isolated from the myometrium (the middle layer of the uterine wall) and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
During pregnancy, hormones and growth factors cause an extensive hypertrophy of uterine smooth muscle cells. This and other morphological and biochemical effects on the uterus can be studied in vitro using HUtSMC.
Ordering information: Human Uterine Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.