VWR® C-Leg Bench Frame with Top and Double Bay Uprights
Supplier: VWR
14-C101VWR415 14-C100VWR414 14-C100VWR416 14-C101VWR452 14-C101VWR416 14-C103VWR452 14-C100VWR454 14-C103VWR454 14-C101VWR413 14-C100VWR413 14-C103VWR414 14-C101VWR455 14-C103VWR413 14-C103VWR453 14-C101VWR414 14-C103VWR416 14-C101VWR454 14-C100VWR453 14-C103VWR455 14-C100VWR455 14-C100VWR415 14-C100VWR452 14-C101VWR453 14-C103VWR415
89524-928EA 7717.38 CAD
89524-928 89524-228 89524-998 89524-922 89524-856 89524-858 89525-000 89524-924 89524-926 89524-286 89524-362 89524-860 89524-222 89524-288 89524-364 89524-224 89524-994 89524-862 89524-358 89524-996 89524-226 89524-290 89524-292 89524-360
VWR® C-Leg Bench Frame with Top and Double Bay Uprights
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Ordering information: Configurations include frame, top, and double bay uprights. Frame color is light neutral. Top includes black phenolic or black laminate options.