VWR® C-Leg C-Leg Bench Frame with Top and Single Bay Uprights
Supplier: VWR
14-C101VWR395 14-C101VWR398 14-C100VWR397 14-C100VWR395 14-C103VWR449 14-C101VWR451 14-C103VWR450 14-C103VWR447 14-C100VWR448 14-C100VWR447 14-C103VWR446 14-C101VWR447 14-C103VWR397 14-C100VWR394 14-C100VWR450 14-C103VWR393 14-C103VWR451 14-C101VWR448 14-C100VWR449 14-C100VWR393 14-C103VWR394 14-C101VWR397 14-C101VWR393 14-C103VWR396 14-C101VWR394 14-C103VWR448 14-C101VWR446 14-C103VWR398 14-C101VWR450 14-C100VWR446 14-C100VWR451 14-C103VWR395 14-C101VWR449 14-C100VWR398 14-C101VWR396 14-C100VWR396
89524-844EA 4460.7 CAD
89524-844 89524-986 89524-920 89524-348 89524-846 89524-208 89524-988 89524-848 89524-352 89524-990 89524-274 89524-354 89524-210 89524-276 89524-278 89524-212 89524-850 89524-852 89524-214 89524-350 89524-918 89524-910 89524-216 89524-854 89524-912 89524-218 89524-914 89524-916 89524-980 89524-982 89524-344 89524-346 89524-984 89524-280 89524-282 89524-284
VWR® C-Leg C-Leg Bench Frame with Top and Single Bay Uprights
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Ordering information: Configurations include frame, top, and single bay uprights. Frame color is light neutral. Top includes black phenolic or black laminate options.