Mesenchymal Stem Cell Medium, PromoCell
Supplier: Promocell
PromoCell Mesenchymal Stem Cell Media were developed for the in vitro expansion and directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue
- Supports the expansion of multipotent human Mesenchymal Stem Cells (MSC)
- In vitro expansion from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue
- Five media to efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages
The PromoCell Mesenchymal Stem Cell Growth Medium supports the expansion of multipotent human Mesenchymal Stem Cells (MSC) without inducing early senescence and differentiation as observed in standard culture media. PromoCell offers five media to efficiently induce differentiation of MSC into adipogenic, chondrogenic, osteogenic or neurogenic lineages, respectively. Note: The PromoCell MSC Chondrogenic Differentiation Medium w/o Inducers (C-28014) must be supplemented with adequate chondrogenic inducers by the customer. Recommended for Human Mesenchymal Stem Cells from Bone Marrow (hMSC-BM), Human Mesenchymal Stem Cells from Umbilical Cord Matrix (hMSC-UC), Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT).
Ordering information: PromoCell Mesenchymal Stem Cell Media are available as Medium (Readyto- use) and consist of a bottle of Basal Medium and one vial of Supplement- Mix. Adding the SupplementMix to the Basal Medium results in the complete Medium.