Human saphenous vein endothelial cells (HSaVEC), PromoCell
Supplier: Promocell
C-12231 C-14025
10171-948EA 2195.44 CAD
10171-948 10171-950 10172-190
Human saphenous vein endothelial cells (HSaVEC), PromoCell
Cells
Primary Human Saphenous Vein Endothelial Cells (HSaVEC) are isolated from human saphe-nous veins from single donors
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
The saphenous vein is often used for autotransplantation in coronary artery bypass operations, when arterial grafts are not available or many grafts are required.
Ordering information: Human Saphenous Vein Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.