Human pulmonary artery endothelial cells (HPAEC), PromoCell
Supplier: Promocell
C-12241 C-14024
10171-952EA 2020.88 CAD
10171-952 10171-954 10172-188
Human pulmonary artery endothelial cells (HPAEC), PromoCell
Cells
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches)
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.
Ordering information: Human Pulmonary Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Order Now
SPECIFICATIONS
Environmentally Preferable