Melanocyte Growth Medium, PromoCell
Supplier: Promocell
The PromoCell Melanocyte Growth Medium is a serum-free, PMA (Phorbol Myristate Acetate) containing medium optimized for the cultivation of human melanocytes
Since PMA is a tumor promoting mitogen, it may interfere with some experimental approaches. In these cases, we recommend using Melanocyte Growth Medium M2. The medium is optimized for primary human cells but can also be used for horse, porcine, murine, and rat melanocytes
Ordering information: PromoCell Melanocyte Growth Medium is available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.