Cryo-SFM Freezing Medium, PromoCell
The PromoCell Freezing Medium Cryo-SFM is an animal component-free, chemically defined medium with DMSO, methylcellulose, and other cryoprotective components developed for the cryopreservation of human and animal primary cells
Note Each freeze/thaw cycle leads to a loss in proliferation potential and cell number, particularly when working with fragile primary cells. If freezing the cells, please do so at as low a passage number as possible.
Recommended for Human Primary Cells, Animal Primary Cells, Established Cell Lines, Induced Pluripotent Stem Cells, Embryonic Stem Cells
It ensures high cell viability, excellent growth rates, and good attachment of the cells after thawing. The medium is the first choice for all types of primary cells and cell lines.