Human aortic endothelial cells (HAoEC), PromoCell
Supplier: Promocell
C-12271 C-14023
10171-970EA 1998.1 CAD
10171-970 10171-972 10172-186
Human aortic endothelial cells (HAoEC), PromoCell
Cells
Primary Human Aortic Endothelial Cells (HAoEC) are isolated from the human ascending (thoracic) and descending (abdominal) aorta
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
They are useful for studying vascular diseases such as thrombosis, atherosclerosis, and hypertension as well as for stent-graft compatibility testing.
Ordering information: Human Umbilical Aortic Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.