Airway Epithelial Cell Growth Medium, PromoCell
Supplier: Promocell
C-21265 C-21060 C-21160 C-21260
10172-270EA 254.21 CAD
10172-270 10175-240 10175-246 10175-254
Airway Epithelial Cell Growth Medium, PromoCell
Cell Culture Media
The PromoCell Airway Epithelial Cell Growth Medium is a serum-free medium optimized for the cultivation of epithelial cells from large air passages
Recommended for Human Nasal Epithelial Cells (HNEpC), Human Bronchial Epithelial Cells (HBEpC), Human Tracheal Epithelial Cells (HTEpC).
The medium is optimized for primary human cells but can also be used for murine, porcine, equine, and rat airway epithelial cells.
Ordering information: PromoCell Airway Epithelial Cell Growth Medium is available as Medium (Readyto- use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately. Airway Epithelial Cell Media.
Order Now
SPECIFICATIONS
Environmentally Preferable