Human umbilical artery endothelial cells (HUAEC), PromoCell
Supplier: Promocell
C-12202 C-14013
10171-908EA 1151.14 CAD
10171-908 10171-958 10175-202
Human umbilical artery endothelial cells (HUAEC), PromoCell
Cells
Primary Human Umbilical Artery Endothelial Cells (HUAEC) are isolated from the arteries of the umbilical cord and are often used for in vitro studies on atherosclerosis but also for endothelial cell-dependent lymphocyte activation and chemoattractant activity experiments
  • Von Willebrand factor (vWF) positive
  • CD31 positive, Dil-Ac-LDL uptake positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
(Human Umbilical Vein Endothelial Cells (HUVEC) from the same donor are available on request).
Ordering information: Human Umbilical Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.