Follicle Dermal Papilla Cell Growth Medium, PromoCell
Supplier: Promocell
C-26500 C-26501 C-26502 C-26505
10172-330EA 230.27 CAD
10172-330 10172-332 10172-334 10172-336
Follicle Dermal Papilla Cell Growth Medium, PromoCell
Cell Culture Media
The PromoCell Follicle Dermal Papilla Cell Growth Medium is a low-serum (4% V/V) medium
The combination of low serum and growth factors leads to excellent proliferation of human follicle dermal papilla cells in vitro.
Ordering information: PromoCell Follicle Dermal Papilla Cell Growth Medium is available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.