Chondrocyte Growth Medium, PromoCell
Supplier: Promocell
The PromoCell Chondrocyte Growth Medium is designed for the optimal cultivation of human chondrocytes
The fetal calf serum in the medium is strictly pretested for optimal performance. The medium is optimized for primary human cells but can also be used for primary porcine and rat chondrocytes.
Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix can also be purchased separately.