Smooth Muscle Cell Growth Medium 2, PromoCell
Supplier: Promocell
The PromoCell Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures
Recommended for Human Umbilical Artery Smooth Muscle Cells (HUASMC), Human Coronary Artery Smooth Muscle Cells (HCASMC), Human Pulmonary Artery Smooth Muscle Cells (HPASMC), Human Aortic Smooth Muscle Cells (HAoSMC), Human Bronchial Smooth Muscle Cells (HBSMC), Human Tracheal Smooth Muscle Cells (HTSMC), Human Bladder Smooth Muscle Cells (HBdSMC), Human Prostate Smooth Muscle Cells (HPrSMC), Human Uterine Smooth Muscle Cells (HUtSMC)
The medium is optimized for primary human cells but can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.
Ordering information: PromoCell Smooth Muscle Cell Media are available as Medium (Ready-to-use) or as Medium Kit. The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.