Myocyte Growth Medium, PromoCell
Supplier: Promocell
C-22270 C-22275 C-22070 C-22170
10172-040EA 251.17 CAD
10172-040 10172-042 10172-286 10175-260
Myocyte Growth Medium, PromoCell
Cell Culture Media
The PromoCell Myocyte Growth Medium is a low serum (5% V/V) medium supplemented with well chosen hormones and growth factors
PromoCell Myocyte Growth Medium is available as Medium (Ready-to-use) or as Medium Kit.
It was developed for the optimal expansion and maintenance of primary cardiac myocytes in culture. The medium is optimized for primary human cells but can also be used for primary porcine cardiac myocytes.
Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.