Normal Human Epidermal Keratinocytes (NHEK), PromoCell
Supplier: Promocell
C-12001 C-12003 C-12005 C-12006 C-14001 C-14002 C-14003 C-14004
10171-886EA 1250.44 CAD
10171-886 10171-888 10171-890 10171-892 10171-894 10171-896 10171-898 10171-900 10172-164 10172-166 10172-168 10172-170
Normal Human Epidermal Keratinocytes (NHEK), PromoCell
Cells
Primary Normal Human Epidermal Keratinocytes (NHEK) are isolated from the epidermis of juvenile foreskin or adult skin from different locations like the face, the breasts, the abdomen, and the thighs. They are the major cell type in the epidermis, making up about 90% of the cells. Epidermal keratinocytes originate in the stratum basale and move up through the layers of the epidermis. During this movement, they undergo gradual differentiation and morphology changes until they reach the stratum corneum, where they form a layer of nucleus-free, flat, and highly keratinized squamous cells. This layer forms an effective barrier to the entry of infectious agents into the body and minimizes moisture loss.
  • Cytokeratin positive
  • Available from juvenile foreskin or adult skin from different locations
  • Available from single and pooled donors
Keratinocytes are also able to produce a variety of cytokines, growth factors, interleukins and complement factors
This product can be used in the following applications. Wound healing research, Epidermal development and differentiation studies, Dermatological research, Drug uptake studies, Pharmaceutical testing, Cosmetic and toxicological testing, Oncology.
Therefore keratinocytes are important for wound healing, inflammation, and immune response. Normal Human Dermal Fibroblasts (NHDF) and Normal Human Epidermal Melanocytes (NHEM) cultured in M2 medium from the same donor are available on request.
Ordering information: Human Epidermal Keratinocytes can be ordered as cryopreserved or proliferating cells and from single or pooled donors. Additional cell pellets stored in RNAlater® are available.