Skeletal Muscle Cell Growth Medium, PromoCell
Supplier: Promocell
C-23060 C-23061 C-23160 C-23260 C-23265
10172-050EA 427.14 CAD
10172-050 10172-052 10172-060 10172-074 10172-076
Skeletal Muscle Cell Growth Medium, PromoCell
Cell Culture Media
The PromoCell Skeletal Muscle Cell Growth Medium is a low-serum (5% V/V) medium optimized for the expansion of human skeletal muscle cells
The unique Skeletal Muscle Cell Differentiation Medium is a serum-free medium that induces the fusion of myoblasts to myotubes with typical multinucleated syncytia. The media are optimized for primary human cells but can also be used for rat, murine, and rabbit skeletal muscle cells.
Ordering information: The Medium (Ready-to-use) consists of a 500 ml bottle of Basal Medium and one vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the Supplements to the Basal Medium results in the complete Growth or Differentiation Medium, respectively. Basal Medium (with or without phenol red) as well as SupplementMix (Growth and Differentiation Medium) and SupplementPack (Growth Medium) can also be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable