Human Bronchial Smooth Muscle Cells (HBSMC), PromoCell
Supplier: Promocell
C-12561 C-14055
10172-100EA 1810.7 CAD
10172-100 10172-102 10172-228
Human Bronchial Smooth Muscle Cells (HBSMC), PromoCell
Cells
Primary Human Bronchial Smooth Muscle Cells (HBSMC) are isolated from the bronchi from single donors and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
They connect the endings of the bronchial cartilage and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the bronchi walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HBSMC are suitable for in vitro studies of asthma and other pulmonary diseases.
Ordering information: Human Bronchial Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.