Human Mononuclear Cells from Peripheral Blood (hMNC-PB), PromoCell
Supplier: Promocell
C-12907 C-14098
10175-178EA 987.03 CAD
10175-178 10175-218
Human Mononuclear Cells from Peripheral Blood (hMNC-PB), PromoCell
Cells
PromoCell Mononuclear Cells (MNC) represent the lymphocyte and monocyte fraction isolated from umbilical cord blood or peripheral blood. They are prepared by optimized low-density gradient centrifugation, which effectively reduces the granulocyte content. Red blood cells are gently but thoroughly depleted by proprietary techniques instead of osmotic lysis. This results in ultra-pure mononuclear cells with superior viability and unchanged biological function. In addition, the cells do not clump after thawing, allowing for easy handling and reproducible results.
The MNC can be used directly for experiments or as a starting material for the isolation of several cell types, e.g., hematopoietic and endothelial progenitors, monocytes, and lymphocyte subpopulations
This product can be used in the following applications. Hematologic and immunologic studies, Oncology research, Virology and infection biology
Adult peripheral blood contains large numbers of mature immune cells but also some progenitor cells. Typical application fields are immunology, oncology, hematology, virology, and infection biology.
Ordering information: Human Mononuclear Cells from Peripheral Blood (hMNC-PB) are available from single or pooled donors