Human Umbilical Artery Smooth Muscle Cells (HUASMC), PromoCell
Supplier: Promocell
C-12500 C-14050
10172-026EA 1162.37 CAD
10172-026 10172-098 10172-220
Human Umbilical Artery Smooth Muscle Cells (HUASMC), PromoCell
Cells
Primary Human Umbilical Artery Smooth Muscle Cells (HUASMC) are isolated from the arteries of the umbilical cord and stain positive for smooth muscle α-actin
  • Smooth muscle alpha-actin positive
  • CD90 negative
  • Routinely morphology tested
  • Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
Physiological and biochemical processes of vascular diseases can be analyzed with HUASMC in culture.
Ordering information: Human Umbilical Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.